Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001172681.1 | 5472 | Intron | NP_001166152.1 | ||
NM_001172682.1 | 5472 | Intron | NP_001166153.1 | ||
NM_152320.2 | 5472 | Intron | NP_689533.2 | ||
XM_005268637.3 | 5472 | UTR 3 | XP_005268694.1 | ||
XM_005268638.3 | 5472 | UTR 3 | XP_005268695.1 | ||
XM_005268639.2 | 5472 | UTR 3 | XP_005268696.1 | ||
XM_005268640.4 | 5472 | Intron | XP_005268697.1 | ||
XM_011537897.1 | 5472 | UTR 3 | XP_011536199.1 | ||
XM_011537898.1 | 5472 | UTR 3 | XP_011536200.1 | ||
XM_017018797.1 | 5472 | UTR 3 | XP_016874286.1 | ||
XM_017018798.1 | 5472 | UTR 3 | XP_016874287.1 | ||
XM_017018799.1 | 5472 | Intron | XP_016874288.1 |