Product Details

SNP ID
rs2072350
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:38380542 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCAGTTAGGGCGGGCAGCACAGTT[C/T]GGGCAAAGGTGTACAGGGATGAGCC
Phenotype
MIM: 613452 MIM: 609966 MIM: 609293
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CATSPERG PubMed Links

Gene Details

Gene
CATSPERG
Gene Name
cation channel sperm associated auxiliary subunit gamma
There are no transcripts associated with this gene.

Gene
GGN
Gene Name
gametogenetin
There are no transcripts associated with this gene.

Gene
PSMD8
Gene Name
proteasome 26S subunit, non-ATPase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002812.4 Intron NP_002803.2
Gene
SPRED3
Gene Name
sprouty related EVH1 domain containing 3
There are no transcripts associated with this gene.

View Full Product Details