Product Details

SNP ID
rs2072594
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4846729 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGCCATCGGTTGTAAAGGAGAA[C/G]TCCCCCCGCCGTTTCTTCGCGGGGC
Phenotype
MIM: 610660 MIM: 609771
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GLYR1 PubMed Links

Gene Details

Gene
GLYR1
Gene Name
glyoxylate reductase 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308096.1 65 Intron NP_001295025.1
NM_001324096.1 65 Intron NP_001311025.1
NM_001324097.1 65 Intron NP_001311026.1
NM_001324098.1 65 Intron NP_001311027.1
NM_032569.3 65 Intron NP_115958.2
XM_005255639.4 65 Intron XP_005255696.2
XM_011522716.2 65 Intron XP_011521018.1
XM_011522717.1 65 Intron XP_011521019.1
XM_017023786.1 65 Intron XP_016879275.1
XM_017023787.1 65 Intron XP_016879276.1
XM_017023788.1 65 Intron XP_016879277.1
XM_017023789.1 65 Intron XP_016879278.1
Gene
UBN1
Gene Name
ubinuclein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079514.2 65 Intron NP_001072982.1
NM_001288656.1 65 Intron NP_001275585.1
XM_005255277.3 65 Intron XP_005255334.1
XM_005255278.3 65 UTR 5 XP_005255335.1
XM_005255279.3 65 Intron XP_005255336.1
XM_011522465.1 65 Intron XP_011520767.1
XM_011522466.2 65 Intron XP_011520768.1
XM_011522467.1 65 Intron XP_011520769.1
XM_017023189.1 65 Intron XP_016878678.1
XM_017023190.1 65 Intron XP_016878679.1
XM_017023191.1 65 Intron XP_016878680.1
XM_017023192.1 65 Intron XP_016878681.1

View Full Product Details