Product Details

SNP ID
rs2240949
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128382625 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGCGAGAGAGGAAGCCTGGCACGC[A/G]CAGTTGCACATCACTTATTCCTGCC
Phenotype
MIM: 612693
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR219A2 PubMed Links

Gene Details

Gene
MIR219A2
Gene Name
microRNA 219a-2
There are no transcripts associated with this gene.

Gene
MIR219B
Gene Name
microRNA 219b
There are no transcripts associated with this gene.

Gene
URM1
Gene Name
ubiquitin related modifier 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135947.2 Intron NP_001129419.1
NM_001265582.1 Intron NP_001252511.1
NM_030914.3 Intron NP_112176.1

View Full Product Details