Product Details

SNP ID
rs2281023
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27309886 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCCTTGGGATAGTGGGAAAGGT[G/T]GCCTTGAGCTTGATCCCAAGGGACT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC105375107 PubMed Links

Gene Details

Gene
LOC105375107
Gene Name
uncharacterized LOC105375107
There are no transcripts associated with this gene.

Gene
POM121L2
Gene Name
POM121 transmembrane nucleoporin like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033482.3 2347 Missense Mutation NP_258443.2

View Full Product Details