Product Details

SNP ID
rs2959535
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:40533896 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAATAACACATGAAGAAATCAAAT[G/T]ACTTTTACAATATGTTCTCGCTATG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C15orf57 PubMed Links

Gene Details

Gene
C15orf57
Gene Name
chromosome 15 open reading frame 57
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080791.2 Intron NP_001074260.1
NM_001080792.2 Intron NP_001074261.1
NM_001289132.1 Intron NP_001276061.1
NM_052849.3 Intron NP_443081.1
Gene
MRPL42P5
Gene Name
mitochondrial ribosomal protein L42 pseudogene 5
There are no transcripts associated with this gene.

View Full Product Details