Product Details

SNP ID
rs2281780
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:43023859 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTCGCTCACGGTCTGTGCCCTAGC[G/T]CTCACTTGTTCAGTGGAGAGACCGC
Phenotype
MIM: 615339
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC15 PubMed Links

Gene Details

Gene
DNAJC15
Gene Name
DnaJ heat shock protein family (Hsp40) member C15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013238.2 Intron NP_037370.2

View Full Product Details