Product Details

SNP ID
rs2282298
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:152511769 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
CCACCACAGGCCCCGCCAGTCCCTC[C/T]GACGCCGACCTCAGAGTTCCAGCTG
Phenotype
MIM: 612619
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CRCT1 PubMed Links

Gene Details

Gene
CRCT1
Gene Name
cysteine rich C-terminal 1
There are no transcripts associated with this gene.

Gene
LCE5A
Gene Name
late cornified envelope 5A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178438.4 411 Nonsense Mutation CGA,TGA R79* NP_848525.1

View Full Product Details