Product Details

SNP ID
rs2288667
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:34882852 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAAAGCAAAAAGAACCAGTGTTTAT[C/T]GAAACAGAGTTCTCCAATCCTAAAG
Phenotype
MIM: 612491
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APIP PubMed Links

Gene Details

Gene
APIP
Gene Name
APAF1 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015957.3 Intron NP_057041.2
XM_011520154.2 Intron XP_011518456.1
XM_017017875.1 Intron XP_016873364.1

View Full Product Details