Product Details

SNP ID
rs2275768
Assay Type
Functionally tested
NCBI dbSNP Submissions
24
Location
Chr.1:215567710 on Build GRCh38
Set Membership
JSNP
Context Sequence [VIC/FAM]
GATGGCGGGAGGGCACTGCGGCAGC[G/T]TCCCCGCGGCGGCGGCCGGCAGCGG
Phenotype
MIM: 613272
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KCTD3 PubMed Links

Gene Details

Gene
KCTD3
Gene Name
potassium channel tetramerization domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001319294.1 526 Missense Mutation GTC,TTC V9F NP_001306223.1
NM_001319295.1 526 UTR 5 NP_001306224.1
NM_016121.4 526 Missense Mutation GTC,TTC V9F NP_057205.2
XM_005273158.1 526 UTR 5 XP_005273215.1

View Full Product Details