Product Details

SNP ID
rs2275976
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:23743803 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTTTGTGTGGGCTCCTCTTTCTA[C/T]AGCTTCTCCCCAGAATGAATCTTCT
Phenotype
MIM: 605017 MIM: 194544
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
VPREB3 PubMed Links

Gene Details

Gene
VPREB3
Gene Name
pre-B lymphocyte 3
There are no transcripts associated with this gene.

Gene
ZNF70
Gene Name
zinc finger protein 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021916.3 1799 Silent Mutation CTA,CTG L446L NP_068735.1

View Full Product Details