Product Details

SNP ID
rs2289488
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:2871725 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGGCTCCTCACGGGGGGCGGGGG[C/G]GCATCCCTCCATGGGACAGGGAGGG
Phenotype
MIM: 610980
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
KCNQ1DN PubMed Links

Gene Details

Gene
KCNQ1DN
Gene Name
KCNQ1 downstream neighbor (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details