Product Details

SNP ID
rs2276977
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:163494778 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TACCTGGGTCTAGAGTGCGGAGCTG[C/T]TCCGTGGCCACGAGGACGTCACCAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMA16 PubMed Links

Gene Details

Gene
TMA16
Gene Name
translation machinery associated 16 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018352.2 258 UTR 5 NP_060822.2

View Full Product Details