Product Details

SNP ID
rs2289970
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:175923865 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTAGGATGAGACAAATAATATAT[A/G]GAGTGATAAAATTTAATAAGGTTTT
Phenotype
MIM: 610236
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LNPK PubMed Links

Gene Details

Gene
LNPK
Gene Name
lunapark, ER junction formation factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305008.1 7182 Intron NP_001291937.1
NM_001305009.1 7182 Intron NP_001291938.1
NM_001305010.1 7182 Intron NP_001291939.1
NM_001305011.1 7182 Intron NP_001291940.1
NM_030650.2 7182 Intron NP_085153.1
XM_005246878.3 7182 UTR 3 XP_005246935.1
XM_005246879.4 7182 UTR 3 XP_005246936.1
XM_005246880.3 7182 UTR 3 XP_005246937.1
XM_006712783.2 7182 UTR 3 XP_006712846.1
XM_011511945.2 7182 UTR 3 XP_011510247.1
XM_017005055.1 7182 UTR 3 XP_016860544.1

View Full Product Details