Product Details

SNP ID
rs2277256
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:58388174 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAAAATTGTGGCATCTTTTTTCTTT[C/T]TATTTCATTCCATAGATGCAAAAAC
Phenotype
MIM: 600438
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TFAM PubMed Links

Gene Details

Gene
TFAM
Gene Name
transcription factor A, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270782.1 Intron NP_001257711.1
NM_003201.2 Intron NP_003192.1
XM_011540120.2 Intron XP_011538422.1
XM_011540121.2 Intron XP_011538423.1

View Full Product Details