Product Details

SNP ID
rs2277381
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:51912243 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCCCCAAGCTCTCAGTCACTTAAA[C/G]ATTGCTCTCCACCCTTCACCTCTAA
Phenotype
MIM: 601284
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ACVRL1 PubMed Links

Gene Details

Gene
ACVRL1
Gene Name
activin A receptor like type 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000020.2 Intron NP_000011.2
NM_001077401.1 Intron NP_001070869.1
XM_005269235.2 Intron XP_005269292.1
XM_011539008.1 Intron XP_011537310.1

View Full Product Details