Product Details

SNP ID
rs2545817
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:11814340 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATCTCGGCTTTCTTGCTTCGAGAG[A/G]GACTAGGTGCCTCCACCAGAGCTTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF440 PubMed Links

Gene Details

Gene
ZNF440
Gene Name
zinc finger protein 440
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152357.2 54 UTR 5 NP_689570.2
XM_005259731.3 54 UTR 5 XP_005259788.1
XM_017026253.1 54 Intron XP_016881742.1
XM_017026254.1 54 UTR 5 XP_016881743.1
Gene
ZNF491
Gene Name
zinc finger protein 491
There are no transcripts associated with this gene.

View Full Product Details