Product Details
- SNP ID
-
rs2243876
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:189752917 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GACACAACAAAGTGCACTGTGTTGC[A/G]TTTGTTAAAATTTCCAGAGCTCTGC
- Phenotype
-
MIM: 609803
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANKAR
PubMed Links
Gene Details
- Gene
- ANKAR
- Gene Name
- ankyrin and armadillo repeat containing
- Gene
- OSGEPL1
- Gene Name
- O-sialoglycoprotein endopeptidase-like 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022353.2 |
1143 |
Silent Mutation |
AAC,AAT |
N342N |
NP_071748.2 |
XM_005246766.3 |
1143 |
Silent Mutation |
AAC,AAT |
N342N |
XP_005246823.1 |
XM_006712685.1 |
1143 |
Silent Mutation |
AAC,AAT |
N342N |
XP_006712748.1 |
XM_006712686.1 |
1143 |
Silent Mutation |
AAC,AAT |
N195N |
XP_006712749.1 |
XM_011511631.1 |
1143 |
Silent Mutation |
AAC,AAT |
N342N |
XP_011509933.1 |
XM_017004676.1 |
1143 |
Silent Mutation |
AAC,AAT |
N342N |
XP_016860165.1 |
XM_017004677.1 |
1143 |
Silent Mutation |
AAC,AAT |
N342N |
XP_016860166.1 |
XM_017004678.1 |
1143 |
Silent Mutation |
AAC,AAT |
N342N |
XP_016860167.1 |
XM_017004679.1 |
1143 |
Silent Mutation |
AAC,AAT |
N342N |
XP_016860168.1 |
XM_017004680.1 |
1143 |
Silent Mutation |
AAC,AAT |
N342N |
XP_016860169.1 |
XM_017004681.1 |
1143 |
Silent Mutation |
AAC,AAT |
N195N |
XP_016860170.1 |
XM_017004682.1 |
1143 |
Silent Mutation |
AAC,AAT |
N195N |
XP_016860171.1 |
XM_017004683.1 |
1143 |
Silent Mutation |
AAC,AAT |
N195N |
XP_016860172.1 |
XM_017004684.1 |
1143 |
Silent Mutation |
AAC,AAT |
N195N |
XP_016860173.1 |
XM_017004685.1 |
1143 |
Silent Mutation |
AAC,AAT |
N195N |
XP_016860174.1 |
- Gene
- OSGEPL1-AS1
- Gene Name
- OSGEPL1 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details