Product Details

SNP ID
rs2586710
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:20599547 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTCAAACCAAGGGCACCTTGCTT[G/T]GCACTGACGCAAACCCGGCCTTTCC
Phenotype
MIM: 604159
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SLC6A5 PubMed Links

Gene Details

Gene
SLC6A5
Gene Name
solute carrier family 6 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318369.1 148 UTR 5 NP_001305298.1
NM_004211.4 148 UTR 5 NP_004202.3
XM_017018544.1 148 Intron XP_016874033.1
XM_017018545.1 148 UTR 5 XP_016874034.1

View Full Product Details