Product Details

SNP ID
rs2564875
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:112249352 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTTAATCAGTGCAAATATGATCCA[C/G]AATGACACAATGTTATTTTCTTCTT
Phenotype
MIM: 611904
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PLET1 PubMed Links

Gene Details

Gene
PLET1
Gene Name
placenta expressed transcript 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145024.1 Intron NP_001138496.1

View Full Product Details