Product Details

SNP ID
rs2233154
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:40775441 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTTGGGACTGGTTTAGGGCGGGGA[C/T]AAGACCAAGAACACAAGTTTCCTTG
Phenotype
MIM: 601340 MIM: 612945
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIA PubMed Links

Gene Details

Gene
MIA
Gene Name
melanoma inhibitory activity
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202553.1 65 Intron NP_001189482.1
NM_006533.3 65 UTR 5 NP_006524.1
Gene
MIA-RAB4B
Gene Name
MIA-RAB4B readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
RAB4B
Gene Name
RAB4B, member RAS oncogene family
There are no transcripts associated with this gene.

Gene
RAB4B-EGLN2
Gene Name
RAB4B-EGLN2 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details