Product Details

SNP ID
rs2745336
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:127119740 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACCTAGCGCAACCGCCTCGCGC[C/G]GGGAGACCTCGGCTGGGCCCTTAGG
Phenotype
MIM: 610574
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RSPO3 PubMed Links

Gene Details

Gene
RSPO3
Gene Name
R-spondin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032784.4 312 Intron NP_116173.2
XM_017011378.1 312 Intron XP_016866867.1
XM_017011379.1 312 UTR 5 XP_016866868.1

View Full Product Details