Product Details

SNP ID
rs2721195
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144451628 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTGCTCTGAGCTGGCCGCCTGGTG[C/T]GGGCTGAGAGTAGAGGGATGGAGCT
Phenotype
MIM: 616635 MIM: 604546
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYHR1 PubMed Links

Gene Details

Gene
CYHR1
Gene Name
cysteine and histidine rich 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001129888.1 Intron NP_001123360.1
NM_032687.3 Intron NP_116076.1
NM_138496.1 Intron NP_612505.1
XM_006716560.3 Intron XP_006716623.1
XM_006716561.3 Intron XP_006716624.1
XM_006716562.3 Intron XP_006716625.2
XM_011517051.1 Intron XP_011515353.1
XM_017013463.1 Intron XP_016868952.1
XM_017013464.1 Intron XP_016868953.1
XM_017013465.1 Intron XP_016868954.1
XM_017013466.1 Intron XP_016868955.1
Gene
TONSL
Gene Name
tonsoku-like, DNA repair protein
There are no transcripts associated with this gene.

View Full Product Details