Product Details

SNP ID
rs2738819
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:75541794 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATTCCAAGTTTAAATAAATAAAA[A/G]TAACTTGGATCCCAATGTGTTTCTA
Phenotype
MIM: 604817 MIM: 614949
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CHST5 PubMed Links

Gene Details

Gene
CHST5
Gene Name
carbohydrate sulfotransferase 5
There are no transcripts associated with this gene.

Gene
TMEM231
Gene Name
transmembrane protein 231
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077416.2 Intron NP_001070884.2
NM_001077418.2 Intron NP_001070886.1

View Full Product Details