Product Details

SNP ID
rs2762927
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:54146427 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGGTGTTGGCACAATCCAGAGGAC[A/C]GCTACCGGTCTCCAGTAGATTTCAC
Phenotype
MIM: 126065
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CYP24A1 PubMed Links

Gene Details

Gene
CYP24A1
Gene Name
cytochrome P450 family 24 subfamily A member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000782.4 2002 Intron NP_000773.2
NM_001128915.1 2002 Intron NP_001122387.1
XM_005260304.4 2002 Intron XP_005260361.1
XM_017027691.1 2002 Intron XP_016883180.1
XM_017027692.1 2002 UTR 3 XP_016883181.1
XM_017027693.1 2002 Intron XP_016883182.1

View Full Product Details