Product Details
- SNP ID
-
rs2782828
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
40
- Location
-
Chr.1:207323654 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- AACAACTTAAATCAATAGAAGCTAC[G/T]GTAAATACAGAAATTGGCACAAATA
- Phenotype
-
MIM: 125240
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CD55
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs28371598] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CD55
- Gene Name
- CD55 molecule (Cromer blood group)
View Full Product Details