Product Details

SNP ID
rs2783234
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:57994501 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGAATCAAAATCATAGCTAAAAA[C/T]CAATGCTATATAGTTTCTATGACAT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C14orf37 PubMed Links

Gene Details

Gene
C14orf37
Gene Name
chromosome 14 open reading frame 37
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001872.3 12638 Intron NP_001001872.2
NM_001320173.1 12638 Intron NP_001307102.1
XM_011536471.2 12638 Intron XP_011534773.1
XM_017021029.1 12638 UTR 3 XP_016876518.1
XM_017021030.1 12638 UTR 3 XP_016876519.1

View Full Product Details