Product Details

SNP ID
rs2233261
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:110913117 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTACCTCCTCCTTGGAAAACCTCTC[C/T]GCCTGCGTGGTCAGCATTTCCCGAA
Phenotype
MIM: 160781
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCDC63 PubMed Links

Gene Details

Gene
CCDC63
Gene Name
coiled-coil domain containing 63
There are no transcripts associated with this gene.

Gene
MYL2
Gene Name
myosin light chain 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000432.3 452 Silent Mutation GCA,GCG A127A NP_000423.2

View Full Product Details