Product Details

SNP ID
rs2233314
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74515654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGCTGCGCTCTCGCCCTTCTCTG[C/G]GACGCGCCGCATAGGCCACCCCTAC
Phenotype
MIM: 610231 MIM: 604240
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DQX1 PubMed Links

Gene Details

Gene
DQX1
Gene Name
DEAQ-box RNA dependent ATPase 1
There are no transcripts associated with this gene.

Gene
PCGF1
Gene Name
polycomb group ring finger 1
There are no transcripts associated with this gene.

Gene
TLX2
Gene Name
T-cell leukemia homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016170.4 760 Missense Mutation GCG,GGG A141G NP_057254.1

View Full Product Details