Product Details

SNP ID
rs2233460
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:67175104 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCCGCCCATGCTGGATAGGACCTG[A/G]GATGCTGCTGGAGCTGAATCGGATG
Phenotype
MIM: 614117 MIM: 602438 MIM: 605235
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EXOC3L1 PubMed Links

Gene Details

Gene
EXOC3L1
Gene Name
exocyst complex component 3 like 1
There are no transcripts associated with this gene.

Gene
HSF4
Gene Name
heat shock transcription factor 4
There are no transcripts associated with this gene.

Gene
KIAA0895L
Gene Name
KIAA0895 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040715.1 869 Intron NP_001035805.1
Gene
NOL3
Gene Name
nucleolar protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185057.2 869 UTR 3 NP_001171986.1
NM_001276307.1 869 UTR 3 NP_001263236.1
NM_001276309.1 869 UTR 3 NP_001263238.1
NM_001276311.1 869 UTR 3 NP_001263240.1
NM_001276312.1 869 UTR 3 NP_001263241.1
NM_001276319.1 869 UTR 3 NP_001263248.1
NM_003946.6 869 UTR 3 NP_003937.1
XM_005256217.2 869 UTR 3 XP_005256274.1
XM_005256219.3 869 UTR 3 XP_005256276.1
XM_011523424.2 869 Nonsense Mutation TGA,TGG *223W XP_011521726.1
XM_017023843.1 869 UTR 3 XP_016879332.1

View Full Product Details