Product Details

SNP ID
rs2233601
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:141489662 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGCCACCCCTGAGCGAGAGATGC[A/G]CATCTCAGAATCAGCAGCATCTGGG
Phenotype
MIM: 606288 MIM: 606297 MIM: 606298 MIM: 603059 MIM: 606289 MIM: 606290 MIM: 606291 MIM: 606292 MIM: 606293 MIM: 606294 MIM: 606295 MIM: 606296 MIM: 606299 MIM: 606300 MIM: 606301 MIM: 603058 MIM: 606302 MIM: 606303 MIM: 606304 MIM: 603627 MIM: 606305 MIM: 606306
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCDHGA1 PubMed Links

Gene Details

Gene
PCDHGA1
Gene Name
protocadherin gamma subfamily A, 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018912.2 422 Intron NP_061735.1
NM_031993.1 422 Intron NP_114382.1
Gene
PCDHGA10
Gene Name
protocadherin gamma subfamily A, 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018913.2 422 Intron NP_061736.1
NM_032090.1 422 Intron NP_114479.1
Gene
PCDHGA11
Gene Name
protocadherin gamma subfamily A, 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018914.2 422 Intron NP_061737.1
NM_032091.1 422 Intron NP_114480.1
NM_032092.1 422 Intron NP_114481.1
Gene
PCDHGA12
Gene Name
protocadherin gamma subfamily A, 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003735.2 422 Intron NP_003726.1
NM_032094.1 422 Intron NP_115265.1
Gene
PCDHGA2
Gene Name
protocadherin gamma subfamily A, 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018915.3 422 Intron NP_061738.1
NM_032009.2 422 Intron NP_114398.1
Gene
PCDHGA3
Gene Name
protocadherin gamma subfamily A, 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018916.3 422 Intron NP_061739.2
NM_032011.1 422 Intron NP_114400.1
Gene
PCDHGA4
Gene Name
protocadherin gamma subfamily A, 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018917.3 422 Intron NP_061740.2
NM_032053.2 422 Intron NP_114442.2
Gene
PCDHGA5
Gene Name
protocadherin gamma subfamily A, 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018918.2 422 Intron NP_061741.1
NM_032054.1 422 Intron NP_114443.1
Gene
PCDHGA6
Gene Name
protocadherin gamma subfamily A, 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018919.2 422 Intron NP_061742.1
NM_032086.1 422 Intron NP_114475.1
Gene
PCDHGA7
Gene Name
protocadherin gamma subfamily A, 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018920.3 422 Intron NP_061743.1
NM_032087.2 422 Intron NP_114476.1
Gene
PCDHGA8
Gene Name
protocadherin gamma subfamily A, 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014004.2 422 Intron NP_054723.1
NM_032088.1 422 Intron NP_114477.1
Gene
PCDHGA9
Gene Name
protocadherin gamma subfamily A, 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018921.2 422 Intron NP_061744.1
NM_032089.1 422 Intron NP_114478.1
Gene
PCDHGB1
Gene Name
protocadherin gamma subfamily B, 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018922.2 422 Intron NP_061745.1
NM_032095.1 422 Intron NP_115266.1
Gene
PCDHGB2
Gene Name
protocadherin gamma subfamily B, 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018923.2 422 Intron NP_061746.1
NM_032096.1 422 Intron NP_115267.1
Gene
PCDHGB3
Gene Name
protocadherin gamma subfamily B, 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018924.4 422 Intron NP_061747.2
NM_032097.2 422 Intron NP_115268.2
Gene
PCDHGB4
Gene Name
protocadherin gamma subfamily B, 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003736.2 422 Intron NP_003727.1
NM_032098.1 422 Intron NP_115269.1
Gene
PCDHGB5
Gene Name
protocadherin gamma subfamily B, 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018925.2 422 Intron NP_061748.1
NM_032099.1 422 Intron NP_115270.1
Gene
PCDHGB6
Gene Name
protocadherin gamma subfamily B, 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018926.2 422 Intron NP_061749.1
NM_032100.1 422 Intron NP_115271.1
Gene
PCDHGB7
Gene Name
protocadherin gamma subfamily B, 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018927.3 422 Intron NP_061750.1
NM_032101.2 422 Intron NP_115272.1
Gene
PCDHGC3
Gene Name
protocadherin gamma subfamily C, 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002588.3 422 Intron NP_002579.2
NM_032402.1 422 Intron NP_115778.1
NM_032403.2 422 Intron NP_115779.1
Gene
PCDHGC4
Gene Name
protocadherin gamma subfamily C, 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018928.2 422 Intron NP_061751.1
NM_032406.1 422 Intron NP_115782.1
Gene
PCDHGC5
Gene Name
protocadherin gamma subfamily C, 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018929.2 422 Missense Mutation CAC,CGC H141R NP_061752.1
NM_032407.1 422 Missense Mutation CAC,CGC H141R NP_115783.1

View Full Product Details