Product Details

SNP ID
rs2270763
Assay Type
Validated
NCBI dbSNP Submissions
82
Location
Chr.1:197905819 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
AGGCTGGCCAGCTAAACTATGTGGA[C/T]CCAGCTACTGGCTATGTGGTGCTCA
Phenotype
MIM: 606066
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf53 PubMed Links

Gene Details

Gene
C1orf53
Gene Name
chromosome 1 open reading frame 53
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024594.2 386 Silent Mutation GAC,GAT D96D NP_001019765.1
Gene
LHX9
Gene Name
LIM homeobox 9
There are no transcripts associated with this gene.

View Full Product Details