Product Details

SNP ID
rs2271053
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:108044018 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAAAAAAATCCCTCAACCAAATAA[A/G]ATACAATAAAAAATAAACGGTTGCC
Phenotype
MIM: 601028
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD47 PubMed Links

Gene Details

Gene
CD47
Gene Name
CD47 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001777.3 4422 UTR 3 NP_001768.1
NM_198793.2 4422 UTR 3 NP_942088.1
XM_005247908.1 4422 UTR 3 XP_005247965.1
XM_005247909.1 4422 UTR 3 XP_005247966.1
XM_017007536.1 4422 Intron XP_016863025.1

View Full Product Details