Product Details
- SNP ID
-
rs2973588
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:126789517 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACATATTTGCCCAGGTTATCCTACC[C/T]AAAAGTTCTTTCCCTACATGGGCTG
- Phenotype
-
MIM: 150340
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LMNB1
PubMed Links
Gene Details
- Gene
- LMNB1
- Gene Name
- lamin B1
View Full Product Details