Product Details
- SNP ID
-
rs2233384
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:233032704 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCCTTCCTGTCCTGCAGAAGGAGAG[A/C]GTGTTCCAGTCGGGAGCCCATGCCT
- Phenotype
-
MIM: 605528
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
NEU2
PubMed Links
Gene Details
- Gene
- NEU2
- Gene Name
- neuraminidase 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005383.2 |
33 |
Missense Mutation |
AGA,AGC |
R11S |
NP_005374.2 |
View Full Product Details