Product Details

SNP ID
rs2291855
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57016407 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTACCTGTGGAGCAGCTCTGTGCC[A/G]GGGGCTGGGTGGTCTGGCAGGATCA
Phenotype
MIM: 162330
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAC3 PubMed Links

Gene Details

Gene
TAC3
Gene Name
tachykinin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178054.1 154 UTR 5 NP_001171525.1
NM_013251.3 154 UTR 5 NP_037383.1
Gene
ZBTB39
Gene Name
zinc finger and BTB domain containing 39
There are no transcripts associated with this gene.

View Full Product Details