Product Details

SNP ID
rs2292111
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:39828599 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAATGAAGTTAACAAACCTAAAGT[G/T]TAAACTGTAGTTGGCATATACTAGG
Phenotype
MIM: 613200
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PDS5A PubMed Links

Gene Details

Gene
PDS5A
Gene Name
PDS5 cohesin associated factor A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100399.1 Intron NP_001093869.1
NM_001100400.1 Intron NP_001093870.1
XM_011513672.2 Intron XP_011511974.1
XM_017007928.1 Intron XP_016863417.1

View Full Product Details