Product Details

SNP ID
rs2292151
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:4816707 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGCCGCCGCCTGCATCCGCTCACC[A/G]TCCAGGTGTTGGCTCTGTTCCCGCA
Phenotype
MIM: 607601
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TICAM1 PubMed Links

Gene Details

Gene
TICAM1
Gene Name
toll like receptor adaptor molecule 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182919.3 1939 Silent Mutation GAC,GAT D557D NP_891549.1

View Full Product Details