Product Details

SNP ID
rs2280286
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:73369741 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTACGCAGCTGCTCCAGGGAGTGGG[C/T]CGGAGTCTGTTTGCCCCGCTGGGAC
Phenotype
MIM: 611687 MIM: 611689
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KHDC3L PubMed Links

Gene Details

Gene
KHDC3L
Gene Name
KH domain containing 3 like, subcortical maternal complex member
There are no transcripts associated with this gene.

Gene
OOEP
Gene Name
oocyte expressed protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080507.2 148 Missense Mutation ACC,GCC T18A NP_001073976.1
XM_017010890.1 148 Missense Mutation ACC,GCC T18A XP_016866379.1
Gene
OOEP-AS1
Gene Name
OOEP antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details