Product Details

SNP ID
rs2991959
Assay Type
Functionally Tested
NCBI dbSNP Submissions
15
Location
Chr.1:234376808 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATGCCTTGCTGTGAGTCTTAAATG[C/T]CCGTGTTCATTAGAGCTCCCTTCTC
Phenotype
MIM: 614772
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
COA6 PubMed Links

Gene Details

Gene
COA6
Gene Name
cytochrome c oxidase assembly factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012985.2 Intron NP_001013003.1
NM_001206641.2 Intron NP_001193570.2
NM_001301733.1 Intron NP_001288662.1
Gene
LOC101927765
Gene Name
uncharacterized LOC101927765
There are no transcripts associated with this gene.

View Full Product Details