Product Details

SNP ID
rs2991979
Assay Type
Functionally tested
NCBI dbSNP Submissions
59
Location
Chr.1:45590186 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTAATTCATCTTTTTTTCCAACT[A/G]TACAAGTACTGCCTTAGTTCACCCT
Phenotype
MIM: 603185
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NASP PubMed Links

Gene Details

Gene
NASP
Gene Name
nuclear autoantigenic sperm protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195193.1 Intron NP_001182122.1
NM_002482.3 Intron NP_002473.2
NM_152298.3 Intron NP_689511.2
XM_005270888.2 Intron XP_005270945.1
XM_005270889.2 Intron XP_005270946.1
XM_011541509.1 Intron XP_011539811.1
XM_017001356.1 Intron XP_016856845.1

View Full Product Details