Product Details

SNP ID
rs2303634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:108517185 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAAACCGGACAGGGCGCAGCATGC[C/T]GCCACCAAGTGCACCAGCCCGTCCA
Phenotype
MIM: 611684
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FICD PubMed Links

Gene Details

Gene
FICD
Gene Name
FIC domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007076.2 359 Silent Mutation GCC,GCT A71A NP_009007.2
Gene
SART3
Gene Name
squamous cell carcinoma antigen recognized by T-cells 3
There are no transcripts associated with this gene.

View Full Product Details