Product Details
- SNP ID
-
rs2303714
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:77077416 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGCGCTCGCCCTGCTCCACGGCCA[C/G]CTCGCGCCGCTCCAGCTCCCGCTCC
- Phenotype
-
MIM: 615250
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SNORA47
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2303713] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SNORA47
- Gene Name
- small nucleolar RNA, H/ACA box 47
There are no transcripts associated with this gene.
- Gene
- ZBED3
- Gene Name
- zinc finger BED-type containing 3
There are no transcripts associated with this gene.
- Gene
- ZBED3-AS1
- Gene Name
- ZBED3 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details