Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001040668.1 | 113 | UTR 5 | NP_001035758.1 | ||
NM_001282516.1 | 113 | UTR 5 | NP_001269445.1 | ||
NM_001282517.1 | 113 | UTR 5 | NP_001269446.1 | ||
NM_001282519.1 | 113 | UTR 5 | NP_001269448.1 | ||
NM_001282520.1 | 113 | UTR 5 | NP_001269449.1 | ||
NM_001282521.1 | 113 | UTR 5 | NP_001269450.1 | ||
NM_138639.1 | 113 | UTR 5 | NP_619580.1 | ||
XM_017027345.1 | 113 | UTR 5 | XP_016882834.1 | ||
XM_017027346.1 | 113 | UTR 5 | XP_016882835.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001197122.1 | 113 | UTR 5 | NP_001184051.1 | ||
NM_001197123.1 | 113 | UTR 5 | NP_001184052.1 | ||
NM_001197124.1 | 113 | UTR 5 | NP_001184053.1 | ||
NM_001197125.1 | 113 | UTR 5 | NP_001184054.1 | ||
NM_001197126.1 | 113 | UTR 5 | NP_001184055.1 | ||
NM_001197127.1 | 113 | UTR 5 | NP_001184056.1 | ||
NM_001197128.1 | 113 | UTR 5 | NP_001184057.1 | ||
NM_001571.5 | 113 | UTR 5 | NP_001562.1 | ||
XM_006723197.1 | 113 | UTR 5 | XP_006723260.1 | ||
XM_006723198.1 | 113 | UTR 5 | XP_006723261.1 | ||
XM_006723200.1 | 113 | Intron | XP_006723263.1 | ||
XM_006723201.1 | 113 | UTR 5 | XP_006723264.1 | ||
XM_006723202.2 | 113 | UTR 5 | XP_006723265.1 | ||
XM_017026766.1 | 113 | UTR 5 | XP_016882255.1 | ||
XM_017026767.1 | 113 | UTR 5 | XP_016882256.1 |