Product Details

SNP ID
rs2304927
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:77088760 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGGTGGTCGCCGACTCGCAGTCC[A/G]CAGGATGACTCAGGGCAGCCCTGAC
Phenotype
MIM: 601504
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
MIR6516 PubMed Links

Gene Details

Gene
MIR6516
Gene Name
microRNA 6516
There are no transcripts associated with this gene.

Gene
SCARNA16
Gene Name
small Cajal body-specific RNA 16
There are no transcripts associated with this gene.

Gene
SEC14L1
Gene Name
SEC14 like lipid binding 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039573.2 118 Intron NP_001034662.2
NM_001143998.1 118 Intron NP_001137470.1
NM_001143999.1 118 Intron NP_001137471.1
NM_001144001.1 118 Intron NP_001137473.1
NM_001204408.1 118 UTR 5 NP_001191337.1
NM_001204410.1 118 Intron NP_001191339.1
NM_003003.3 118 Intron NP_002994.3
Gene
SNHG20
Gene Name
small nucleolar RNA host gene 20
There are no transcripts associated with this gene.

View Full Product Details