Product Details

SNP ID
rs3012659
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:72346811 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGGAGGGCTACAAAGCCTATGTC[A/G]GGGGCCAGAATTTCAAAGGTTGCCC
Phenotype
MIM: 300269
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HDAC8 PubMed Links

Gene Details

Gene
HDAC8
Gene Name
histone deacetylase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166418.1 Intron NP_001159890.1
NM_001166419.1 Intron NP_001159891.1
NM_001166420.1 Intron NP_001159892.1
NM_001166422.1 Intron NP_001159894.1
NM_001166448.1 Intron NP_001159920.1
NM_018486.2 Intron NP_060956.1
XM_011530986.2 Intron XP_011529288.2
XM_011530987.2 Intron XP_011529289.2
XM_017029640.1 Intron XP_016885129.1
XM_017029641.1 Intron XP_016885130.1
XM_017029642.1 Intron XP_016885131.1
XM_017029643.1 Intron XP_016885132.1
XM_017029644.1 Intron XP_016885133.1
XM_017029645.1 Intron XP_016885134.1
XM_017029646.1 Intron XP_016885135.1
XM_017029647.1 Intron XP_016885136.1

View Full Product Details