Product Details

SNP ID
rs3020786
Assay Type
Functionally Tested
NCBI dbSNP Submissions
1
Location
Chr.1:155291558 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGAGTGTGTTGCTAAGTAGCACG[C/G]CCGTCTGCCTGTGTGGCTATGCTGA
Phenotype
MIM: 609973 MIM: 609712
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
HCN3 PubMed Links

Gene Details

Gene
HCN3
Gene Name
hyperpolarization activated cyclic nucleotide gated potassium channel 3
There are no transcripts associated with this gene.

Gene
PKLR
Gene Name
pyruvate kinase, liver and RBC
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000298.5 Intron NP_000289.1
NM_181871.3 Intron NP_870986.1
XM_006711386.3 Intron XP_006711449.1
XM_011509640.2 Intron XP_011507942.1
XM_017001493.1 Intron XP_016856982.1

View Full Product Details