Product Details
- SNP ID
-
rs2355655
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:50007813 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAGGGAGCTGGGGCTGCCCTTGGAC[A/G]CCTCAGGCACAGGCCCTCATCCTCT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LINC01588
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs116820185] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LINC01588
- Gene Name
- long intergenic non-protein coding RNA 1588
There are no transcripts associated with this gene.
- Gene
- LINC01599
- Gene Name
- long intergenic non-protein coding RNA 1599
There are no transcripts associated with this gene.
View Full Product Details