Product Details

SNP ID
rs2393109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:116857624 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGGGAGGGAGAGTGATATTTTATC[A/C]CTGTCAGAAACATAATAATAGGAGA
Phenotype
MIM: 603447
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HRK PubMed Links

Gene Details

Gene
HRK
Gene Name
harakiri, BCL2 interacting protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003806.2 4603 Intron NP_003797.1
XM_017020146.1 4603 UTR 3 XP_016875635.1
Gene
RNFT2
Gene Name
ring finger protein, transmembrane 2
There are no transcripts associated with this gene.

View Full Product Details