Product Details

SNP ID
rs7487682
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:48185882 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCCTTTGTGTATTCCTTCTCAAT[G/T]CTTTCCTTGGTGTTAACCTTAATAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCDC184 PubMed Links

Gene Details

Gene
CCDC184
Gene Name
coiled-coil domain containing 184
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013635.3 2300 UTR 3 NP_001013657.3

View Full Product Details